Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta I Tubulin Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Monkey, Clone: 1C3J0, Novus Biologicals™
SDP

Catalog No. NB166951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166951 100 μg
NB166952 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166951 Supplier Novus Biologicals Supplier No. NBP315928100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

beta I Tubulin Monoclonal antibody specifically detects beta I Tubulin in Human, Mouse, Rat, Bovine, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen beta I Tubulin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C3J0
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 352-451 of human beta I Tubulin (Q9H4B7).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 81027
Target Species Human, Mouse, Rat, Bovine, Canine, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.