Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta II Tubulin B Antibody (5B2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP174037UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174037UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP174037UV Supplier Novus Biologicals Supplier No. NBP174037UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta II Tubulin B
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5B2
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog
Gene Symbols TUBB2B
Host Species Mouse
Immunogen Recombinant human beta-Tubulin (1-445aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 347733
Test Specificity Anti-human beta-Tubulin mAb, clone 5B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human beta-Tubulin protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.