Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta II Tubulin B Antibody (OTI6D11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272505A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272505A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272505A700 Supplier Novus Biologicals Supplier No. NBP272505AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta II Tubulin B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6D11
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog
Gene Symbols TUBB2B
Host Species Mouse
Immunogen Full length recombinant protein of human beta II Tubulin B (NP_821080) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 347733.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.