Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-III Tubulin Antibody (AA10), Novus Biologicals™
SDP

Catalog No. NBP130048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130048 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130048 Supplier Novus Biologicals Supplier No. NBP130048
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen beta-III Tubulin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone AA10
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:10-1:500
Gene Accession No. Q4QRB4
Gene Alias beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acid residues specific to beta III tubulin conjugated to KLH. Accession # Q4QRB4
Molecular Weight of Antigen 55 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity Specific for the ∼55kDa beta III tubulin protein. This clone is similar to the monoclonal antibody Tuj1.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.