Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-III Tubulin Antibody (CL5813), Novus Biologicals™
SDP

Catalog No. NB403031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB403031 25 μL
NBP261430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB403031 Supplier Novus Biologicals Supplier No. NBP26143025UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen beta-III Tubulin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5813
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:50000 - 1:100000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50000 - 1:100000
Formulation PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
Gene Alias beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids: YEDDEEESEAQGPK
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.