Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-III Tubulin Antibody (TUBB3/3731), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB380554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380554 Supplier Novus Biologicals Supplier No. NBP308439AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen beta-III Tubulin
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone TUBB3/3731
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Host Species Mouse
Immunogen A synthetic peptide (aa 437-450) of human Tubulin beta 3 protein (TUBB3); coupled to KLH. (Uniprot: Q13509 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10381
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.