Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Lactoglobulin B Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062383 0.5 mL
NB017466 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062383 Supplier Novus Biologicals Supplier No. NB10062383
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Beta Lactoglobulin B Polyclonal specifically detects Beta Lactoglobulin B in Bovine samples. It is validated for ELISA.

Specifications

Antigen Beta Lactoglobulin B
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P02754
Gene Alias beta-lactoglobulin pseudogene
Gene Symbols LOC138159
Host Species Sheep
Immunogen Beta-lactoglobulin B from milk
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138159
Test Specificity This product recognizes bovine beta-lactoglobulin B, a component of whey thought to be involved in retinol transport. This antibody also demonstrates some cross reactivity with beta-lactoglobulin A.
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.