Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Antibody (D03), Alexa Fluor™ 647
SDP

Catalog No. NB109206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109206 Supplier Novus Biologicals Supplier No. NBP289768AF647

Rabbit Monoclonal Antibody

beta-NGF Monoclonal specifically detects beta-NGF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen beta-NGF
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D03
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human beta-NGF (NP_002497.2; Ser122-Arg239).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.