Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 532, Clone: 25623, Novus Biologicals™
SDP

Catalog No. N1201013A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1201013A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1201013A32 Supplier Novus Biologicals Supplier No. NB12010513AF532

Mouse Monoclonal Antibody

beta-NGF Monoclonal specifically detects beta-NGF in Human, Mouse, Rat samples. It is validated for ELISA.

Specifications

Antigen beta-NGF
Applications ELISA
Classification Monoclonal
Clone 25623
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Gene Symbols NGF
Host Species Mouse
Immunogen Recombinant full length protein (Human).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4803.0
Test Specificity NGF beta (25623.1)
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.