Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Rat anti-Mouse, Clone: NGF, Novus Biologicals™
SDP

Catalog No. NBP24242225 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24242225 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24242225 Supplier Novus Biologicals Supplier No. NBP24242225UL

Rat Monoclonal Antibody

beta-NGF Monoclonal specifically detects beta-NGF in Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.

Specifications

Antigen beta-NGF
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone NGF
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100, ELISA, Immunofluorescence
Formulation PBS with no preservative
Gene Accession No. P01139
Gene Alias Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Gene Symbols NGF
Host Species Rat
Immunogen Full length for beta-NGF protein was used as the immunogen for this antibody.
Quantity 25 μL
Primary or Secondary Primary
Gene ID (Entrez) 4803
Reconstitution Reconstitute with 0.1 mL water.
Target Species Mouse
Content And Storage Lyophilized antibody can be kept at 4°C for up to 3 months and should be kept at -20°C for long-term storage. To avoid freeze-thaw cycles, reconstituted antibody should be aliquoted before freezing for short-term storage (-20°C) or for long-term storage (-80°C). For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.