Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-TrCP1/BTRC Antibody (OTI2H2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP271402A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271402A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271402A94 Supplier Novus Biologicals Supplier No. NBP271402AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-TrCP1/BTRC Monoclonal specifically detects beta-TrCP1/BTRC in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta-TrCP1/BTRC
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2H2
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias beta-transducin repeat containing, betaTrCP, BETA-TRCP, beta-TrCP1, bTrCP, bTrCP1, E3RSIkappaB, F-box and WD repeats protein beta-TrCP, F-box/WD repeat-containing protein 1A, FBW1A, FBXW1, FBXW1ABTRCP, Fwd1, MGC4643, pIkappaBalpha-E3 receptor subunit
Gene Symbols BTRC
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 52-354 of human beta-TrCP1/BTRC (NP_378663) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8945
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.