Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Tubulin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527552 Supplier Invitrogen™ Supplier No. PA527552
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, NIH-3T3, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TUBB2A
Gene Accession No. Q13885, Q7TMM9, Q9BVA1, Q9CWF2
Gene Alias 2410129E14Rik; 2t; b2 tubulin; B2t; bA506K6.1; beta 2 tubulin; BETA 85D; beta tubulin; beta(2)Tu; beta(2)Tub; beta(2)Tubulin; beta;Tub85D; beta[[2]]-tubulin; BETA2; beta2 tubulin; beta2t; beta2tub; beta2-tub; beta2tubulin; Beta-2-tubulin; beta2-tubulin; beta85D; betaTub; beta-tub; betaTub2; BetaTub85D; beta-tub85D; betaTub85D-PA; beta-Tubulin; beta-Tubulin 85D; beta-Tubulin at 85D; beta-tubulin T beta15 (aa 1-445); betaTubulin85D; beta-tubulin85D; brdp; CDCBM5; CG9359; CG9359-PA; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; D.m.BETA-85D; dJ40E16.7; Dmbeta2; Dmel\CG9359; Dmel_CG9359; DTB4; GH02051p; M(beta)2; ms(3)KK[D]; PMGYSA; RGD1309427; t beta-15; testis-specific beta-tubulin; Tub; Tub2A; TUBB; TUBB2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TubB85D; tubulin; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; Tubulin beta class IIa; Tubulin beta-2 chain; Tubulin beta-2A chain; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; UniProtKB:Q7TMM9}
Gene Symbols TUBB2A, TUBB2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human beta Tubulin 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22151, 347733, 7280, 73710
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.