Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin Antibody (1B12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB360394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360394 Supplier Novus Biologicals Supplier No. NBP250061AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell Western

Specifications

Antigen beta Tubulin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Single Cell Western
Classification Monoclonal
Clone 1B12
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell Western
Formulation 50mM Sodium Borate
Gene Alias beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Host Species Mouse
Immunogen Pig brain beta Tubulin preparation. [UniProt# Q767L7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203068
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.