Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB60936A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60936A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60936A750 Supplier Novus Biologicals Supplier No. NB600936AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta Tubulin Polyclonal specifically detects beta Tubulin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Chinese Hamster, Insect, Invertebrate, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen beta Tubulin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Gene Symbols TUBB
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human beta Tubulin (within residues 1-100). [Swiss-Prot: P07437]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 203068
Target Species Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Chinese Hamster, Invertebrate, Primate, Xenopus species, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.