Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges, 5μm Particle Size
Click to view available options
Diameter (Metric):
2.1 mm
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
Description
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Specifications
Specifications
| Carbon Load | 7% |
| pH | 2 to 9 |
| Particle Size | 5 μm |
| Length (Metric) | 10 mm |
| Pore Size | 150Å |
| Surface Area | 200 m2/g |
| Diameter (Metric) | 4 mm |
| For Use With (Equipment) | Uniguard Direct-connection and Stand Alone Holder |
| Endcapped | Yes |
| USP Type | L7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction