Learn More
betacellulin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$2175.00
Specifications
Antigen | betacellulin |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-271-8565
![]() |
Invitrogen™
500P254BT500UG |
500 μg |
Each of 1 for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Betacellulin. Anti-Human Betacellulin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBetacellulin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Betacellulin (500-P254) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBetacellulin. Western Blot: To detect hBetacellulin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBetacellulin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P254BT-1MG will be provided as 2 x 500 μg
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.Specifications
betacellulin | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Human | |
P35070 | |
685 | |
E.coli-derived Recombinant Human Betacellulin. | |
Antigen affinity chromatography | |
BTC |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin | |
BTC | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.