Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BETP
GREENER_CHOICE

Catalog No. p-4835931
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of GLP-1 receptors

Selective positive allosteric modulator and partial agonist of the glucagon-like peptide 1 (GLP-1) receptor. Increases binding affinity of oxyntomodulin for the GLP-1 receptor. Potentiates oxyntomodulin-mediated GLP-1 receptor signaling in vitro and insulin secretion in vivo. Has no effect on GLP-2, GIP, PTH or glucagon receptors.

Chemical Identifiers

CAS 1371569-69-5
Molecular Formula C20H17F3N2O2S

Specifications

Quantity 10 mg
Formula Weight 406.42
Molecular Weight (g/mol) 406.423
Chemical Name or Material BETP
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.