Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bevacizumab Recombinant Human Monoclonal Antibody (Bevacizumab)
GREENER_CHOICE

Catalog No. PIMA548044
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548044 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548044 Supplier Invitrogen™ Supplier No. MA548044
Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specificity: This antibody binds native and reduced human VEGF (isoform 165).

Bevacizumab (Avastin) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with colon cancer, lung cancer, glioblastoma, renal-cell carcinoma, and age-related macular degeneration. As angiogenesis inhibitor, Bevacizumab blocks angiogenesis by inhibiting vascular endothelial growth factor A (VEGF-A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bevacizumab
Applications ELISA, Flow Cytometry, Inhibition Assays, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone Bevacizumab
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene MUC1
Gene Accession No. P15941
Gene Alias Avastin
Host Species Human
Immunogen Recombinant human VEGF.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.