Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BFAR Antibody, Novus Biologicals™
SDP

Catalog No. NB10056094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056094 0.05 mL
NB10056094T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056094 Supplier Novus Biologicals Supplier No. NB10056094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

BFAR Polyclonal specifically detects BFAR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen BFAR
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias BARRNF47RING finger protein 47, bifunctional apoptosis inhibitor, bifunctional apoptosis regulator
Gene Symbols BFAR
Host Species Rabbit
Immunogen A recombinant protein corresponding to amino acids 1-139 of human BAR was used as immunogen. Human BAR is 450 amino acid protein; GenBank no. NP_057645.1.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 51283
Test Specificity BAR
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.