Learn More
Description
Specifications
Specifications
| Antigen | BFAR |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | BARRNF47RING finger protein 47, bifunctional apoptosis inhibitor, bifunctional apoptosis regulator |
| Gene Symbols | BFAR |
| Host Species | Rabbit |
| Immunogen | A recombinant protein corresponding to amino acids 1-139 of human BAR was used as immunogen. Human BAR is 450 amino acid protein; GenBank no. NP_057645.1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
