Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BICD1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290892 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290892 Supplier Novus Biologicals Supplier No. NBP178735PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BICD1 Polyclonal antibody specifically detects BICD1 in Human samples. It is validated for Western Blot, Immunoprecipitation, KnockDown

Specifications

Antigen BICD1
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation, Knockdown Validated
Formulation PBS
Gene Alias Bicaudal D (Drosophila) homolog 1, bicaudal D homolog 1 (Drosophila), BICD, Bic-D 1, cytoskeleton-like bicaudal D protein homolog 1, protein bicaudal D homolog 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 550 and 600 of human Bicaudal D Homolog 1 using the numbering given in entry NP_001705.2 (GeneID 636).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 636
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.