Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BICD2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017266 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017266 Supplier Novus Biologicals Supplier No. NBP281955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BICD2 Polyclonal specifically detects BICD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen BICD2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS
Gene Alias bicaudal D homolog 2 (Drosophila), KIAA0699
Gene Symbols BICD2
Host Species Rabbit
Immunogen BICD2 antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human BICD2. The immunogen is located within the last 50 amino acids of BICD2. Amino Acid Squence: AKAAPKTKPATPS
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299
Test Specificity This BICD2 antibody will not cross-react with BICD1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.