Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[3.3.0]octane, Thermo Scientific™

Catalog No. AC468520010
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 694-72-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number 00060882
InChI Key AEBWATHAIVJLTA-UHFFFAOYSA-N
IUPAC Name octahydropentalene
SMILES C1CC2CCCC2C1

Specifications

Alpha Vector BICYCLOOCTANE
Color Colorless
Boiling Point 137°C
Flash Point 26°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Bicyclo[3.3.0]octane
Hazard Category Flame
Hazard Statement Exclamation mark
Recommended Storage Normal conditions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.