Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BID Antibody (005), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB108037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108037 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108037 Supplier Novus Biologicals Supplier No. NBP289516C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BID Monoclonal specifically detects BID in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BID
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 5
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BID (P55957-1; Met1-Asp195).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 637
Test Specificity No cross-reactivity in ELISA with: Mouse BID
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.