Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BID Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB017431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017431 0.025 mg
NB100220601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017431 Supplier Novus Biologicals Supplier No. NB10022060.025MG

Rabbit Polyclonal Antibody

BID Polyclonal specifically detects BID in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen BID
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. P70444
Gene Alias apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID
Gene Symbols BID
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 45 of mouse Bid.
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 637
Test Specificity This detects both full length (22 kDa) and weakly reacts wtih cleaved fragment (15 kDa) of mouse Bid.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.