Learn More
Trajan Scientific and Medical Big Trap - Oxygen
SDP
Catalog No. SG1035318
Click to view available options
Port Type:
1/4 in.
1/8 in.
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
  • High capacity filters ideal for bulk purification - capacity of 3 L oxygen
  • Efficiency < 1 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
Specifications
For Use With (Application) ECD and TCD when used with a moisture filter, PID when used with a hydrocarbon filter, ICP-AES (ICP-OES) and ICP-MS
Model Oxygen
Volume (Metric) 750 cm3
Port Type 1/8 in.
Description Removes oxygen as well as traces of sulfur and chlorine compounds from carrier gas
For Use With Big Trap Mounting Clip
Pressure 250 psig
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.