Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biglycan Antibody, Novus Biologicals™
SDP

Catalog No. NB10055407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055407 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055407 Supplier Novus Biologicals Supplier No. NB10055407
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 4 publications

Biglycan Polyclonal specifically detects Biglycan in Bovine, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Peptide ELISA.

Specifications

Antigen Biglycan
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P21810
Gene Alias biglycan, Bone/cartilage proteoglycan I, bone/cartilage proteoglycan-I, dermatan sulphate proteoglycan I, DSPG1, PGI, PG-S1, SLRR1Abiglycan proteoglycan, small leucine-rich protein 1A
Gene Symbols BGN
Host Species Goat
Immunogen Biglycan Antibody is made to a peptide with sequence C-DRLAIQFGNYKK corresponding to C-Terminus according to NP_001702.1.
Molecular Weight of Antigen 41.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 633
Test Specificity Predicted cross-reactivity based on sequence identity: Mouse, Dog.
Target Species Bovine, Human, Rat, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.