Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIIB 021, Tocris Bioscience™

Catalog No. 460810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-081-0 Supplier R&D Systems Supplier No. 4608/10
This item is not returnable. View return policy

Selective Hsp90 inhibitor

Selective, Hsp90 competitive inhibitor (Ki = 1.7 nM). Displays no significant activity at a range of ATP-binding kinases or Na+K+ ATPase. Induces degradation of HER-2 in vitro (EC50 = 38 nM in MCF-7 cells). Inhibits growth and promotes cell death in a variety of human tumor cells. Orally bioavailable.

Chemical Identifiers

CAS 848695-25-0
Molecular Formula C14H15ClN6O
Molecular Weight (g/mol) 318.765
InChI Key QULDDKSCVCJTPV-UHFFFAOYSA-N
PubChem CID 16736529
IUPAC Name 6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]purin-2-amine
SMILES CC1=CN=C(C(=C1OC)C)CN2C=NC3=C2N=C(N=C3Cl)N

Specifications

Quantity 10 mg
Synonym biib021, 6-chloro-9-4-methoxy-3,5-dimethylpyridin-2-yl methyl-9h-purin-2-amine, biib 021, biib-021, unii-851b9fq7q0, 6-chloro-9-4-methoxy-3,5-dimethylpyridin-2-yl methyl purin-2-amine, 6-chloro-9-4-methoxy-3,5-dimethyl-2-pyridinyl methyl-9h-purin-2-amine, 3qdd
Formula Weight 318.76
Percent Purity >98%
Chemical Name or Material BIIB 021
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.