Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biliverdin Reductase A/BLVRA Antibody, Novus Biologicals™
SDP

Catalog No. NB1203466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203466 Supplier Novus Biologicals Supplier No. NB1203466

Chicken Polyclonal Antibody

Biliverdin Reductase A/BLVRA Polyclonal specifically detects Biliverdin Reductase A/BLVRA in Human samples. It is validated for Western Blot.

Specifications

Antigen Biliverdin Reductase A/BLVRA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24
Gene Symbols BLVRA
Host Species Chicken
Immunogen Purified human biliverdin-IX alpha-reductase expressed in E. coli.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644
Test Specificity NB120-3466 detects biliverdin-IX alpha reductase (BLVRA) from human cells.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.