Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Antibody (3C5), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB173489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173489 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173489 Supplier Novus Biologicals Supplier No. NBP280102AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Bim Monoclonal antibody specifically detects Bim in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Bim
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3C5
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
Host Species Rat
Immunogen Synthetic peptide corresponding to aa 20-40 of mouse BimL.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10018
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.