Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB214294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB214294 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB214294 Supplier Novus Biologicals Supplier No. NBP176963C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bim Polyclonal antibody specifically detects Bim in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Bim
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid peptide corresponding to amino acids near the amino terminus of human Bim. The immunogen is located within the first 50 amino acids of Bim. Amino Acid Squence: AERPPQLRPGAPTSLQTEPQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10018
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.