Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB345435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345435 Supplier Novus Biologicals Supplier No. NBP300198JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bim Polyclonal antibody specifically detects Bim in Human samples. It is validated for ELISA

Specifications

Antigen Bim
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Bim (Accession#: NP_006529.1; Ala2-Arg120).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10018
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.