Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Mouse anti-Human, Mouse, Rat, FITC, Clone: 1C2H4, Novus Biologicals™
SDP

Catalog No. NB206115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206115 Supplier Novus Biologicals Supplier No. NBP175511F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bim Monoclonal antibody specifically detects Bim in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Bim
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 1C2H4
Conjugate FITC
Formulation PBS
Gene Alias BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
Host Species Mouse
Immunogen Antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10018
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.