Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BIMU 8

Click to view available options
Quantity:
5 mg
25 mg
Description
BIMU 8 is a potent 5-HT4 receptor full agonist (KD = 77 nM and EC50 = 18 nM for wild type 5-HT4 receptors). Binds to receptors in a mechanism different to that of the endogenous ligand 5-HT.
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride |
| CAS | 134296-40-5 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C19H26N4O2.HCl |
| Formula Weight | Observed MW: 378.9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction