Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179648
Description
BIN2 Polyclonal specifically detects BIN2 in Human samples. It is validated for Western Blot.Specifications
| BIN2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2 | |
| Rabbit | |
| 62 kDa | |
| 100 μL | |
| Cell Cycle and Replication | |
| 51411 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_057377 | |
| BIN2 | |
| Synthetic peptide directed towards the middle region of human BIN2The immunogen for this antibody is BIN2. Peptide sequence EEVLPSSTTPSPGGALSPSGQPSSSATEVVLRTRTASEGSEQPKKRASIQ. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Pig: 86%; Bovine: 79%; Rabbit: 79%; Rat: 77%. | |
| Human, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction