Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | BIN2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
BIN2 Polyclonal specifically detects BIN2 in Human samples. It is validated for Western Blot.Specifications
| BIN2 | |
| Polyclonal | |
| Rabbit | |
| NP_057377 | |
| 51411 | |
| Synthetic peptide directed towards the middle region of human BIN2The immunogen for this antibody is BIN2. Peptide sequence EEVLPSSTTPSPGGALSPSGQPSSSATEVVLRTRTASEGSEQPKKRASIQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2 | |
| BIN2 | |
| IgG | |
| 62 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title