Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100805
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5100805 100 μL
1 options

Catalog No. PIPA5100805

Supplier: Invitrogen™ PA5100805

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BIN2.

BAR proteins, which are characterized by a common N-terminal BAR domain, are a family of adaptor proteins implicated in a diverse set of cellular processes. BIN1 (bridging integrator 1) and BIN2 are BAR proteins. The human BIN2 gene is located at chromosome 4q22.1, and encodes a 565-amino acid protein with a predicted molecular weight of ~62 kDa. It shows preferential expression in hematopoietic tissues, with highest mRNA levels in spleen and peripheral blood leukocytes and also high levels in thymus, colon, and placenta. BIN2 has been shown to migrate with an apparent mobility of ~80 kDa, and interact with BIN1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BIN2
Gene Accession No. Q9UBW5
Gene Alias BIN2; BRAP1; BRAP-1; breast cancer associated protein BRAP1; breast cancer-associated protein 1; Bridging integrator 2; bridging integrator-2
Gene Symbols BIN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BIN2(Accession Q9UBW5), corresponding to amino acid residues S440-Q490.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51411
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.