Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIN3 Antibody (3A4), Novus Biologicals™
SDP

Catalog No. NBP221676 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221676 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221676 Supplier Novus Biologicals Supplier No. NBP221676
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BIN3 Monoclonal specifically detects BIN3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen BIN3
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3A4
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1500, ELISA 1:5000-1:50000, Immunoprecipitation 10-100 ul
Gene Accession No. Q9NQY0
Gene Alias bridging integrator 3, MGC14978
Gene Symbols BIN3
Host Species Mouse
Immunogen BIN3 Antibody (3A4) was produced in mouse by repeated immunizations with a full length human BIN3 recombinant protein followed by hybridoma development. (Uniprot: Q9NQY0)
Molecular Weight of Antigen 29.66 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55909
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.