Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIN3 Antibody (OTI8E9), PE, Novus Biologicals™
SDP

Catalog No. NBP272051PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272051PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272051PE Supplier Novus Biologicals Supplier No. NBP272051PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BIN3 Monoclonal specifically detects BIN3 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen BIN3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI8E9
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias bridging integrator 3, MGC14978
Gene Symbols BIN3
Host Species Mouse
Immunogen Full length recombinant protein of human BIN3 (NP_061158) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55909.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.