Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Bio-bin™ Waste Disposal Containers
SCI_ED

Catalog No. S29245 Shop All Dynalab Corporation Products
Click to view available options
Dimensions (L x W x H):
11 x 11 x 16.3 in.
16.85 x 7.12 x 11.25 in.
6 x 6 x 15 in.
6.7 x 6.7 x 7.87 in.
7.1 x 10.8 x 12.32 in.
9 x 4.3 x 4.7 in.
Capacity (Metric):
2.5 L
6 L
30 L
30L
Product Type:
Disposal Container
M Stand
U Stand
Z Stand

Bio-bin™ is the ideal solution for biohazard or non-hazardous waste disposal. Reduce cost, weight, transport, CO2 emissions, storage space!

  • Space efficient, lightweight fold-flat design
  • Built-in lid and handles
  • Temporary and permanent seals
  • Stackable post use
  • UN 3291 Approved
  • Printed user instructions and user ID/date information on lid
  • Waterproof construction 

Specifications

Capacity (Metric) 6 L
Certifications/Compliance UN 3291
Dimensions (L x W x H) 7.1 x 10.8 x 12.32 in.
Height (Metric) 312.928 mm
Length (Metric) 180.34 mm
Shape U-shaped
Width (Metric) 274.32 mm
Autoclavable Autoclavable
For Use With (Application) Holding 6L Bio-Bin™ at a 30 degree angle
For Use With (Equipment) 6L Bio-Bin™
Width (English) 10.8 in.
Length (English) 7.1 in.
Height (English) 12.32 in.
Material Stainless Steel
Product Type U Stand
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.