Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio Plas™ Uni-Flex™ Safety Caps
Click to view available options
Color:
Gray
Green
Lavender
Natural
Red
Yellow
For Use With (Equipment):
12 mm Culture or 13 mm Blood Collection Tubes
13 mm Culture Tubes
13 mm Culture and Blood Collection Tubes
16 mm Culture and Blood Collection Tubes
Description
Protects laboratory staff from infectious aerosols
- Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
- Designed to hold under centrifugation, agitation, refrigeration and freezing
- Stretchable, unique, over-locking design to prevent evaporation
- One-hand application and removal
- Trace-metal free material
Specifications
Specifications
Product Type | Safety Cap |
Includes | Liquid-tight seal |
Material | Polyethylene |
Color | Natural |
For Use With (Equipment) | 12 mm Culture or 13 mm Blood Collection Tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction