Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Fast-Trac Activity Wheel, Certified
SDP

Catalog No. p-4185757
Encompass
Click to view available options
Color:
Amber
Blue

Specialized activity wheel for use with the Mouse Igloo shelter (Mfr. Nos. K3328, K3570, and K3327)

  • Supports natural and instinctive running behavior
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)

Specifications

Autoclavable Autoclavable
Product Type Fast-Trac Wheel Activity
Color Amber
Material Polycarbonate
For Use With (Application) Supports natural and instinctive running behavior
For Use With (Equipment) Mouse igloo
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.