Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 18 HPLC Columns

Catalog No. 03254031
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
20 mm
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-254-031 250 mm 20 mm
72105254630 250 mm 4.6 mm
03-050-864 250 mm 2.1 mm
03-050-862 150 mm 1 mm
72105152130 150 mm 2.1 mm
03-050-859 100 mm 2.1 mm
72-105-052130 50 mm 2.1 mm
03-050-858 100 mm 1 mm
72105014001 10 mm 4 mm
03-050-856 10 mm 1 mm
03-050-860 125 mm 3 mm
03-050-861 100 mm 4.6 mm
03-050-857 10 mm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 03-254-031 Supplier Thermo Scientific™ Supplier No. 72105259270A
Add to Cart
Add to Cart
Request a Quote

Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.

Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.

  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY

Specifications

Column Format Preparative Column, Standard
Diameter (Metric) 20 mm
Length (Metric) 250 mm
Max. Pressure 4300 psi (295 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.