Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 18 PEEK Bio-Inert 5μm HPLC Columns

Catalog No. 72105104668
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
72105104668 100 mm 4.6 mm
72105102168 100 mm 2.1 mm
72105152168 150 mm 2.1 mm
72105154668 150 mm 4.6 mm
72105252168 250 mm 2.1 mm
72105254668 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 72105104668 Supplier Thermo Scientific™ Supplier No. 72105104668

Outstanding separation of small to medium peptides.

Thermo Scientific™ BioBasic 18 PEEK Bio-Inert 5μm HPLC Columns offer outstanding separation of small to medium peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules.

  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Particle size: 5μm
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available in bio-inert column hardware. Please call Customer Service for more information.

Recommended for:

Separation of medium to low hydrophobicity peptides.

TRUSTED_SUSTAINABILITY

Specifications

Column Format PEEK Column
Diameter (Metric) 4.6 mm
For Use With LC/MS
Length (Metric) 100 mm
Particle Size 5 μm
pH 2 to 10
Pore Size 300 Å
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.