Learn More
Thermo Scientific™ BioBasic™ 4 HPLC Columns
Description
Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
- Based on 300Å silica for outstanding biomolecule performance
- Lower carbon load for optimal retention of larger peptides and proteins
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 250 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 2 to 9 |
| Pore Size | 300 Å |
| Surface Area | 100 m²/g |
| Temperature | 60°C |
| USP Type | L26 |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.