Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 4 KAPPA 5μm Capillary HPLC Columns

Catalog No. 72305050365
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
Diameter (Metric):
0.18 mm
0.32 mm
0.5 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
72305050365 50 mm 0.32 mm
72305050265 50 mm 0.18 mm
72305050565 50 mm 0.5 mm
72305100565 100 mm 0.5 mm
72305150365 150 mm 0.32 mm
72305150565 150 mm 0.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 72305050365 Supplier Thermo Scientific™ Supplier No. 72305050365

Thermo Scientific™ BioBasic™ 4 KAPPA 5μm Capillary HPLC Columns operate at the low flowrates required for greater ionization efficiencies.

Thermo Scientific™ BioBasic™ 4 KAPPA 5μm Capillary HPLC Columns operate at the low flowrates required for greater ionization efficiencies. Highly efficient KAPPA capillary columns meet the sensitivity needs of demanding LC/MS applications, particularly proteomics.

  • Rugged sleeved design for robustness
  • Excellent for LC/MS analysis
  • Particle size: 5μm

Features of BioBasic 4 Solid Phase

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides an proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY

Specifications

Column Format Capillary Column
Diameter (Metric) 0.32 mm
For Use With LC/MS, APCI, ESI
Length (Metric) 50 mm
Particle Size 5 μm
pH 2 to 10
Pore Size 300 Å
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 4%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.