Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 4 PEEK Bio-Inert HPLC Columns, 5μm Particle Size

Catalog No. 03051717
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-051-717 150 mm 4.6 mm
72305102168 100 mm 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-051-717 Supplier Thermo Scientific™ Supplier No. 72305154668

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Thermo Scientific™ BioBasic 4 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are designed for the analysis of larger proteins and peptides.

Thermo Scientific™ BioBasic 4 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are designed for the analysis of larger proteins and peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other phases and column dimensions are available in bio-inert column hardware. Please call Customer Service for more information

TRUSTED_SUSTAINABILITY

Specifications

Column Format PEEK Column
Diameter (Metric) 4.6 mm
For Use With LC/MS
Length (Metric) 150 mm
Particle Size 5 μm
pH 2 to 10
Pore Size 300 Å
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 4%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.