Learn More
Thermo Scientific™ BioBasic™ 8 KAPPA Guard Columns
Description
Protect your analytical HPLC column from damage caused by sample debris and interfering substances. It's the economical way to extend the lifetime of your analytical column with minimal impact on performance.
Use Thermo Scientific™ BioBasic™ 8 KAPPA Guard Columns with BioBasic 8 KAPPA HPLC Columns, designed to operate at low flow rates required for greater ionization efficiencies and meet the sensitivity needs of demanding LC/MS applications, particularly proteomics.
Features of BetaBasic 8 Solid Phase
- 300Å pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Optimized for LC/MS separations

Specifications
Specifications
Carbon Load | 5% |
pH | 2 to 10 |
Particle Size | 5 μm |
Length (Metric) | 30 mm |
Pore Size | 300Å |
Diameter (Metric) | 0.18 mm |
For Use With (Application) | LC/MS |
Endcapped | Yes |
USP Type | L7 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.