Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Biocytin
GREENER_CHOICE

Catalog No. 334910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-491-0 10 mg
33-495-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-491-0 Supplier Tocris Bioscience™ Supplier No. 3349/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Versatile marker for neuroanatomical investigations

Versatile marker used in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays. Displays high solubility in aqueous solutions and has a low molecular weight facilitating injection using micropipettes. Can be incorporated with a variety of avidin and streptavidin conjugates for detection by light, fluorescence or electron microscope.

Chemical Identifiers

CAS 576-19-2
Molecular Formula C16H28N4O4S

Specifications

Quantity 10 mg
Formula Weight 372.48
Purity >98%
Molecular Weight (g/mol) 372.484
Chemical Name or Material Biocytin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.