Learn More
Therapak™ Biological Substance Category B Replenishment Supplies
For shipment of biological specimens
$159.00 - $359.50
Specifications
Applications For shipment of Biological Substance Category B specimens
View More Specs
Products 5
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
22-130-059
View Documents
Therapak™
37909
Medium Insulated EPS Cooler Medium
Case of 4 for $359.50
Only null left
Add to Cart
22-130-060
View Documents
Therapak™
37910
Large Insulated EPS Cooler Large
Case of 2 for $316.00
Only null left
Add to Cart
22-130-072
View Documents
Therapak™
56400
8 oz. Refrigerant Pack 8 oz. pack
Case of 72 for $159.00
Only null left
Add to Cart
22-130-414
Accessories View Documents
Therapak™
56407
3 oz. Refrigerant Pack 3 oz. pack
Case of 192 for $337.50
Only null left
Add to Cart
22-130-058
View Documents
Therapak™
37908
Small Insulated EPS Cooler Small N/A N/A N/A
Description
Description
  • Insulated Shippers include standard DOT/IATA labels, EPS cooler, and corrugated shipper box
  • Leakproof Refrigerant Shippers keep lab specimens cold (30°F) with nontoxic, food-grade gel refrigerants
Specifications
Specifications
For shipment of Biological Substance Category B specimens
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.